missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fumarase Protein
A cDNA sequence encoding the Fumarase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10621-10ug
Additional Details : Weight : 0.01000kg
Specifications
2271 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 25 unit/mg, and is defined as the amount of enzyme that cleaves 1umole of L-Malate to Fumarate per minute at pH 7.5 at 37C. | |
Human | |
Untagged | |
The Fumarase protein solution (1 mg/ml) contains 20mM Tris-HCl, pH 8. |
Fumarase Protein | |
Research Use Only | |
FH | |
Recombinant Protein | |
E. coli | |
MASQNSFRIE YDTFGELKVP NDKYYGAQTV RSTMNFKIGG VTERMPTPVI KAFGILKRAA AEVNQDYGLD PKIANAIMKA ADEVAEGKLN DHFPLVVWQT GSGTQTNMNV NEVISNRAIE MLGGELGSKI PVHPNDHVNK SQSSNDTFPT AMHIAAAIEV HEVLLPGLQK LHDALDAKSK EFAQIIKIGR THTQDAVPLT LGQEFSGYVQ QVKYAMTRIK AAMPRIYELA AGGTAVGTGL NTRIGFAEKV AAKVAALTGL PFVTAPNKFE ALAAHDALVE LSGAMNTTAC SLMKIANDIR FLGSGPRSGL GELILPENEP GSSIMPGKVN PTQCEAMTMV AAQVMGNHVA VTVGGSNGHF ELNVFKPMMI KNVLHSARLL GDASVSFTEN CVVGIQANTE RINKLMNESL MLVTALNPHI GYDKAAKIAK TAHKNGSTLK ETAIELGYLT AEQFDEWVKP KDMLGPK | |
Greater than 95.0% as determined by SDS-PAGE. |