missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FSH alpha/beta Protein

Product Code. 18309467
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18309467

Brand: R&D Systems™ 5925FS010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human FSH alpha/beta Protein is derived from CHO. The Recombinant Human FSH alpha/beta Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 2488
Molecular Weight (g/mol) 10.2 kDa (FSH alpha) & 12.4 kDa (FSH beta)
Name FSH alpha/beta
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived human FSH alpha/beta protein, (N-terminus) Human FSH alpha Ala25-Ser116 Accession &Num; P01215 Human FSH beta Cys21-Glu129 Accession &Num; P01225 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Noncovalently-linked heterodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.