missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FRZB Protein
A cDNA sequence encoding the FRZB was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | FRZB Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958738
|
enQuireBio™
QP11900-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15948738
|
enQuireBio™
QP11900-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15938738
|
enQuireBio™
QP11900-1mg |
1 mg |
£2975.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
FRZB Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FRZB protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSAACEPVR IPLCKSLPWN MTKMPNHLHH STQANAILAI EQFEGLLGTH CSPDLLFFLC AMYAPICTID FQHEPIKPCK SVCERARQGC EPILIKYRHS WPENLACEEL PVYDRGVCIS PEAIVTADGA DFPMDSSNGN CRGASSERCK CKPIRATQKT YFRNNYNYVI RAKVKEIKTK CHDVTAVVEV KEILKSSLVN IPRDTVNLYT SSGCLCPPLN VNEEYIIMGY EDEERSRLLL VEGSIAEKWK DRLGKKVKRW DMKLRHLGLS KSDSSNSDST QSQKSGRNSN PRQARN | |
Greater than 90.0% as determined by SDS-PAGE. |