missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fractalkine / CX3CL1 Protein
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
Specifications
Specifications
| Accession Number | NP_001291321.1 |
| Gene ID (Entrez) | 6376 |
| Name | Fractalkine / CX3CL1 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | CX3CL1 |
| Biological Activity | Human Fractalkine / CX3CL1 induces Human PBMC chemotaxis (significanty different from control at Human Fractalkine / CX3CL1 concentrations greater than 10-100 ng/ml). |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction