missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fractalkine / CX3CL1 Protein

Product Code. 15925879
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
This item is not returnable. View return policy

Product Code. 15925879

Brand: enQuireBio™ QP5299500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_001291321.1
Gene ID (Entrez) 6376
Name Fractalkine / CX3CL1 Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol CX3CL1
Biological Activity Human Fractalkine / CX3CL1 induces Human PBMC chemotaxis (significanty different from control at Human Fractalkine / CX3CL1 concentrations greater than 10-100 ng/ml).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence QHHGVTKCNI . TCSKMTSKI P. VALLIHYQ QN. QASCGKR AII. LETRQH RLFC. ADPKE QWVKD. AMQH LDRQAA. ALT RNG
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.