missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FOLR2 Protein

Product Code. 18395597
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395597 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395597 Supplier R&D Systems™ Supplier No. 5697FR050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Folic Acid-Bovine Serum Albumin was coated at 5 μg/mL (100 μL/well), the concentration of Recombinant Human FOLR-2 that produces 50% of the optimal binding response was found to be <1 nM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14207
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 2350
Molecular Weight (g/mol) M.W. (Observed): 25-40 kDa, reducing conditions; M.W. (theoretical): 25.4 kDa
Name FOLR2
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human FOLR2 protein Gln22-His228, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol FOLR2
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein FOLR2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.