missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Follistatin Protein

Product Code. 18668737 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18668737 500 μg 500µg
18663507 1 mg 1mg
18610639 100 μg 100µg
18638037 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18668737 Supplier Novus Biologicals™ Supplier No. NBP307105500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2 mm filtered concentrated solution in 20 mM Tris-HCl (pH 8.5), 150 mM NaCl.
Gene ID (Entrez) 10468
Molecular Weight (g/mol) MolecularWeight-theroretical: 31.5 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1 EU/μg of Follistatin as determined by LAL method.
Gene Alias follistatin, follistatin isoform FST317, FSActivin-binding protein
Gene Symbol FST
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Briefly centrifuge vial prior to opening to bring the contents to the bottom. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to 0.1-1.0 mg/mL Stock solutions should be apportioned into working aliquots and stored (at -20°C.)
Form Purified
Research Category Cancer
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein Follistatin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.