missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FLT4 Protein
A cDNA sequence encoding the FLT4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10618-2ug
Additional Details : Weight : 0.01000kg
Specifications
FLT4 Protein | |
Research Use Only | |
Measured by its ability to bind recombinant rat VEGF-C in a functional solid phase binding assay. Immobilised recombinant human VEGFR-3/FLT-4 at 5 microg/ml can bind recombinant rat VEGF-C in a linear range of 8-500 ng/ml. | |
Human | |
His | |
FLT4 was lyophilized from a concentrated (1 mg/ml) sterile solution containing 1xPBS. |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
MQRGAALCLR LWLCLGLLDG LVSGYSMTPP TLNITEESHV IDTGDSLSIS CRGQHPLEWA WPGAQEAPAT GDKDSEDTGV VRDCEGTDAR PYCKVLLLHE VHANDTGSYV CYYKYIKARI EGTTAASSYV FVRDFEQPFI NKPDTLLVNR KDAMWVPCLV SIPGLNVTLR SQSSVLWPDG QEVVWDDRRG MLVSTPLLHD ALYLQCETTW GDQDFLSNPF LVHITGNELY DIQLLPRKSL ELLVGEKLVL NCTVWAEFNS GVTFDWDYPG KQAERGKWVP ERRSQQTHTE LSSILTIHNV SQHDLGSYVC KANNGIQRFR ESTEVIVHEN PFISVEWLKG PILEATAGDE LVKLPVKLAA YPPPEFQWYK DGKALSGRHS PHALVLKEVT EASTGTYTLA LWNSAAGLRR NISLELVVNV PPQIHEKEAS SPSIYSRHSR QALTCTAYGV PLPLSIQWHW RPWTPCKMFA QRSLRRRQQQ DLMPQCRDWR AVTTQDAVNP IESLDTWTEF VEGKNKTVSK LVIQNANVSA MYKCVVSNKV GQDERLIYFY VTTIPDGFTI ESKPSEELLE GQPVLLSCQA DSYKYEHLRW YRLNLSTLHD AHGNPLLLDC KNVHLFATPL AASLEEVAPG ARHATLSLSI PRVAPEHEGH YVCEVQDRRS HDKHCHKKYL SVQALEAPRL TQNLTDLLVN VSDSLEMQCL VAGAHAPSIV WYKDERLLEE KSGVDLADSN QKLSIQRVRE EDAGRYLCSV CNAKGCVNSS ASVAVEGSED KGSMEIVILV GTGVIAVFFW VLLLLIFCNM RRPAHADIKT GYLSIIMDPG EVPLEEQCEY LSYDASQWEF PRERLHLGRV LGYGAFGKVV EASAFGIHKG SSCDTVAVKM LKEGATASEH RALMSELKIL IHIGNHLNVV | |
Greater than 90.0% as determined by SDS-PAGE. |