missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Flt-3 Ligand/FLT3L Protein, Animal-Free
Click to view available options
Quantity:
1 mg
50 μg
Unit Size:
1mg
50µg
Description
Measured in a cell proliferation assay using OCI-AML5 acute myeloid leukemia cells. The ED50 for this effect is 0.200-2.00 ng/mL. The specific activity of Recombinant Human Flt-3 Ligand/FLT3L is >5.00 x 105 units/mg, which is calibrated against the human Flt-3 Ligand WHO Standard (NIBSC code: 96/532).
Specifications
Specifications
| Accession Number | P49771.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 2323 |
| Molecular Weight (g/mol) | M.W. (Observed): 17 kDa, under reducing conditions., M.W. (Theroretical): 18 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived human Flt-3 Ligand/FLT3L protein Thr27 - Ala181 Produced using non-animal reagents in an animal-free laboratory. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | FL, Flt3 ligand, FLT3L, fms-related tyrosine kinase 3 ligand, SL cytokine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction