missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Flt-3 Ligand/FLT3L Protein

Product Code. p-4719681
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Unit Size:
25µg
5µg
This item is not returnable. View return policy

Product Code. 18177080

Brand: R&D Systems™ 308FKN005/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with mouse Flt-3. The ED50 for this effect is 0.2-1 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 2323
Molecular Weight (g/mol) 18 kDa
Name Flt-3 Ligand
Quantity 5 μg
Source Mouse myeloma cell line,NS0-derived human Flt-3 Ligand/FLT3L protein Thr27-Pro185
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute 5 μg vials at 50 μg/mL in sterile PBS. Reconstitute 25 μg or larger vials at 100-250 μg/mL in sterile PBS.
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.