missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Flt-3 Ligand/FLT3L Protein

Product Code. 18775886
Click to view available options
Quantity:
100 μg
250 μg
Unit Size:
100µg
250µg
This item is not returnable. View return policy

Product Code. 18775886

Brand: R&D Systems™ 308FKN100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with mouse Flt-3. The ED50 for this effect is 0.2-1 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAA17999
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 2323
Molecular Weight (g/mol) 18 kDa
Quantity 100 μg
Source Mouse myeloma cell line,NS0-derived human Flt-3 Ligand/FLT3L protein Thr27-Pro185
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Gene Symbol FLT3LG
Conjugate Unconjugated
Cross Reactivity Flt-3 Ligand
Species Human
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.