missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Flt-3/Flk-2 Fc Chimera Protein

Product Code. 18187950
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18187950

Brand: R&D Systems™ 368ST050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Flt-3/Flk-2 Fc Chimera Protein is derived from NS0. The Recombinant Human Flt-3/Flk-2 Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 2322
Molecular Weight (g/mol) 85.5 kDa (monomer)
Name Flt-3/Flk-2 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human Flt-3/Flk-2 protein, (N-terminus) Human Flt-3 (Asn27-Asn541) Accession &Num; AAA18947 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.