missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FKBP1B Protein
A cDNA sequence encoding the FKBP1B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10607-5ug
Additional Details : Weight : 0.01000kg
Specifications
2281 | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 300 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin. | |
Human | |
His | |
The FKBP1B solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT. |
FKBP1B Protein | |
Research Use Only | |
FKBP1B | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH RSMGVEIETI SPGDGRTFPK KGQTCVVHYT GMLQNGKKFD SSRDRNKPFK FRIGKQEVIK GFEEGAAQMS LGQRAKLTCT PDVAYGATGH PGVIPPNATL IFDVELLNLE | |
Greater than 90.0% as determined by SDS-PAGE analysis. |