missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FKBP12 Protein

Product Code. 18158430
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18158430

Brand: R&D Systems™ 3777FK100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human FKBP12 Protein is derived from E. coli. The Recombinant Human FKBP12 Protein has been validated for the following applications: Inhibition Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2μm filtered solution in HEPES and NaCl.
Gene ID (Entrez) 2280
Molecular Weight (g/mol) 13 kDa
Name FKBP12
Quantity 100 μg
Source E. coli-derived human FKBP12 protein Gly2-Glu108,with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.