missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Ficolin-3 Protein
Description
The Recombinant Human Ficolin-3 Protein is derived from NS0. The Recombinant Human Ficolin-3 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and NaCl with Trehalose. |
| Gene ID (Entrez) | 8547 |
| Molecular Weight (g/mol) | 30.7 kDa |
| Name | Ficolin-3 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Ficolin-3 protein Lys22-Arg299,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | FCN3, FCNH, Ficolin3, HAKA1, H-Ficolin |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?