missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Fibromodulin/FMOD Protein

Product Code. 18351678
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351678 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351678 Supplier R&D Systems™ Supplier No. 9840FM050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA.

When Bovine Collagen I is coated at 10 μg/mL (100 μL/well), Recombinant Human Fibromodulin/FMOD (Catalog # 9840-FM) binds with an ED50 of 0.250-2.25 μg/mL.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q06828
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 2331
Molecular Weight (g/mol) 36 kDa
Name Collagen-binding 59 kDa protein, fibromodulin, fibromodulin proteoglycan, FM, FMOD, Keratan sulfate proteoglycan fibromodulin, SLRR2E, SLRR2EKSPG fibromodulin
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human Fibromodulin/FMOD protein Asp75-Ile376,with an N-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 200μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.