missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGFR4 Protein
A cDNA sequence encoding the FGFR4 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 2264 |
---|---|
Name | FGFR4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | FGFR4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15918668
|
enQuireBio™
QP11877-1UG |
1 μg |
£99.15
1µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938668
|
enQuireBio™
QP11877-5UG |
5 μg |
£155.00
5µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928668
|
enQuireBio™
QP11877-50UG |
50 μg |
£1320.00
50µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
2264 | |
Research Use Only | |
FGFR4 | |
Human | |
His | |
FGFR4 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
FGFR4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
LEASEEVELE PCLAPSLEQQ EQELTVALGQ PVRLCCGRAE RGGHWYKEGS RLAPAGRVRG WRGRLEIASF LPEDAGRYLC LARGSMIVLQ NLTLITGDSL TSSNDDEDPK SHRDPSNRHS YPQQAPYWTH PQRMEKKLHA VPAGNTVKFR CPAAGNPTPT IRWLKDGQAF HGENRIGGIR LRHQHWSLVM ESVVPSDRGT YTCLVENAVG SIRYNYLLDV LERSPHRPIL QAGLPANTTA VVGSDVELLC KVYSDAQPHI QWLKHIVING SSFGADGFPY VQVLKTADIN SSEVEVLYLR NVSAEDAGEY TCLAGNSIGL SYQSAWLTVL PEEDPTWTAA APEARYTDLE HHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |