missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF7 / FGF-7 / KGF Protein
Click to view available options
Quantity:
10 μg
100 μg
2 μg
250 μg
Unit Size:
100µg
10µg
250µg
2µg
Specifications
Specifications
| Accession Number | NP_002000.1 |
| Gene ID (Entrez) | 2252 |
| Name | FGF7 / FGF-7 / KGF Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than 1.0 EU/ug as determined by LAL method. |
| Gene Symbol | FGF7 |
| Protein Length | Partial (see sequence information for more details). |
| Biological Activity | The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg. |
| Product Type | Recombinant Protein |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction