missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FGF4 Protein

Product Code. 15915939
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15915939 100 μg 100µg
15935939 500 μg 500µg
15925939 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15915939 Supplier enQuireBio™ Supplier No. QP5343100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_001998.1
Gene ID (Entrez) 2249
Name FGF4 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol FGF4
Biological Activity Recombinant human FGF-4 induced 3T3 cell proliferation was measured at varying concentrations. Bioactivity was confirmed and an ED50 of approximately 3 ng/ml was measured.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MAPTAPNGTL EAELERRWES LVALSLARLP VAAQPKEAAV QSGAGDYLLG IKRLRRLYCN VGIGFHLQAL PDGRIGGAHA DTRDSLLELS PVERGVVSIF GVASRFFVAM SSKGKLYGSP FFTDECTFKE ILLPNNYNAY ESYKYPGMFI ALSKNGKTKK GNRVSPTMKV THFLPRL
Buffer 10 mM sodium phosphate, 75mM sodium chloride, 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.