missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF4 Protein
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Spezifikation
Spezifikation
| Accession Number | NP_001998.1 |
| Gene ID (Entrez) | 2249 |
| Name | FGF4 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | FGF4 |
| Biological Activity | Recombinant human FGF-4 induced 3T3 cell proliferation was measured at varying concentrations. Bioactivity was confirmed and an ED50 of approximately 3 ng/ml was measured. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur