missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF21 / Fibroblast Growth Factor 21 Protein
A DNA sequence encoding the Homo sapiens (Human) FGF21 / Fibroblast Growth Factor 21, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP1070-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_061986.1 | |
FGF21 / Fibroblast Growth Factor 21 Protein | |
Research Use Only | |
FGF21 | |
Recombinant Protein | |
E. coli | |
HPIPDSSPLL QFGGQVRQRY LYTDDAQQTE AHLEIREDGT VGGAADQSPE SLLQLKALKP GVIQILGVKT SRFLCQRPDG ALYGSLHFDP EACSFRELLL EDGYNVYQSE AHGLPLHLPG NKSPHRDPAP RGPARFLPLP GLPPALPEPP GILAPQPPDV GSSDPLSMVG PSQGRSPSYA S | |
Greater than 90% as determined by SDS-PAGE. |
26291 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |