missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF10 / KGF2 Protein
A DNA sequence encoding the Homo sapiens (Human) FGF10 / KGF2 Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6039-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_004456.1 | |
FGF10 / KGF2 Protein | |
Research Use Only | |
FGF10 | |
Recombinant Protein | |
E. coli | |
CQALGQDMVS PEATNSSSSS FSSPSSAGRH VRSYNHLQGD VRWRKLFSFT KYFLKIEKNG KVSGTKKENC PYSILEITSV EIGVVAVKAI NSNYYLAMNK KGKLYGSKEF NNDCKLKERI EENGYNTYAS FNWQHNGRQM YVALNGKGAP RRGQKTRRKN TSAHFLPMVV HS | |
Greater than 80% as determined by SDS-PAGE. |
2255 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |