missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FGF basic/FGF2/bFGF (146 aa) Protein

Product Code. 18759646
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18759646 10 μg 10µg
1 options
This item is not returnable. View return policy

Product Code. 18759646

Brand: R&D Systems™ 233FB010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.1-0.6 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09038
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 2247
Molecular Weight (g/mol) 16.5 kDa
Quantity 10 μg
Source E. coli-derived human FGF basic/FGF2/bFGF protein Pro143-Ser288,with an N-terminal Ala
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Gene Symbol FGF2
Conjugate Unconjugated
Cross Reactivity FGF basic/FGF2
Species Human
Reconstitution Reconstitute at 100-250 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.