missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FGF basic/FGF2/bFGF (146 aa) Protein

Product Code. 18716856
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18716856

Brand: R&D Systems™ 233FB010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.1-0.6 ng/mL.
TRUSTED_SUSTAINABILITY

Specifikationer

Accession Number P09038
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl.
Gene ID (Entrez) 2247
Molecular Weight (g/mol) 16.5 kDa
Quantity 10 μg
Source E. coli-derived human FGF basic/FGF2/bFGF protein Pro143-Ser288,with an N-terminal Ala
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Gene Symbol FGF2
Conjugate Unconjugated
Cross Reactivity FGF basic/FGF2
Species Human
Reconstitution Reconstitute at 100-250 μg/mL in sterile PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.