missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FGF basic/FGF2/bFGF (145 aa) Protein

Product Code. 18739906
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. Quantity unitSize
18739906 10 μg 10µg
18727916 25 μg 25µg
2 options
This item is not returnable. View return policy

Product Code. 18739906

Brand: R&D Systems™ 3718FB010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells.

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.1-0.6 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001997
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl.
Gene ID (Entrez) 2247
Molecular Weight (g/mol) M.W. (theoretical): 16 kDa
Name FGF basic 145 aa
Quantity 10 μg
Source E. coli-derived human FGF basic/FGF2/bFGF protein Ala144-Ser288
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Gene Symbol FGF2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2 um filtered solution in MOPS, Na2SO4, EDTA and DTT.
Endotoxin Level <0.01 EU / 1 ug of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein FGF basic/FGF2/bFGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.