missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human FGF acidic/FGF1 Protein

Product Code. 18623287 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
This item is not returnable. View return policy

Product Code. 18623287

Brand: Novus Biologicals™ NBP261310500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

FGF acidic/FGF1 protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine balb/c 3T3 cells is less than 0.5 ng/mL, corresponding to a specific activity of > 2.0 x 10^6 IU/mg in the presence of 10 μg/mL of heparin.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4, 2 mM EDTA, 0.5 mM DTT.
Gene ID (Entrez) 2246
Molecular Weight (g/mol) MolecularWeight-theroretical: 17.3 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 0.1 EU/μg of FGF acidic/FGF1 as determined by LAL method.
Gene Alias AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1
Gene Symbol FGF1
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Angiogenesis, Biologically Active Proteins, Cancer, Stem Cell Markers
Purity or Quality Grade >97 % pure by SDS-PAGE and HPLC
Protein FGF acidic/FGF1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.