missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FGF-5 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£184.66
Specifications
Gene ID (Entrez) | 2250 |
---|---|
Molecular Weight (g/mol) | 27 kDa |
Name | FGF-5 |
Quantity | 50μg |
Source | E. coli-derived human FGF-5 protein Glu23-Gly268 (Lys238Asn and Pro245Ser),with an N-terminal Met & Leu26-Gly268 (Lys238Asn and Pro245Ser) |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18117290
|
R&D Systems™
237-F5-050 |
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with Trehalose and with BSA as a carrier protein. |
£184.66
50µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18187860
|
R&D Systems™
237-F5-050/CF |
Supplied as a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT. |
£184.66
50µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human FGF-5 Protein is derived from E. coli. The Recombinant Human FGF-5 Protein has been validated for the following applications: Bioactivity.Specifications
2250 | |
FGF-5 | |
E. coli-derived human FGF-5 protein Glu23-Gly268 (Lys238Asn and Pro245Ser),with an N-terminal Met & Leu26-Gly268 (Lys238Asn and Pro245Ser) | |
Recombinant |
27 kDa | |
50μg | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |