missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fibroblast Growth Factor 4 protein is fully biologically active when compared to standard. The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 ng/mL, corresponding to a specific activity of >2.0 x 106IU/mg.
Specifications
Specifications
| Accession Number | P08620, P08620, P08620 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 1 x PBS, 350 mM NaCl, 0.02% Tween-20, pH 7.0. |
| Gene ID (Entrez) | 2249 |
| Molecular Weight (g/mol) | 19.8 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of FGF-4 as determined by LAL method. |
| Gene Alias | fibroblast growth factor 4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?