missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FGF-23 Protein
Description
The Recombinant Human FGF-23 Protein is derived from NS0. The Recombinant Human FGF-23 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT. |
| Gene ID (Entrez) | 8074 |
| Molecular Weight (g/mol) | 26.1 kDa |
| Name | FGF-23 |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived human FGF-23 protein Tyr25-Ile251 (Arg179Gln),with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?