missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF 18 Protein
A cDNA sequence encoding the FGF 18 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10598-1mg
Additional Details : Weight : 0.01000kg
Specifications
FGF 18 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
AEENVDFRIH VENQTRARDD VSRKQLRLYQ LYSRTSGKHI QVLGRRISAR GEDGDKYAQL LVETDTFGSQ VRIKGKETEF YLCMNRKGKL VGKPDGTSKE CVFIEKVLEN NYTALMSAKY SGWYVGFTKK GRPRKGPKTR ENQQDVHFMK RYPKGQPELQ KPFKYTTVTK RSRRIRPTHP A | |
Greater than 95.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
Untagged | |
FGF-18 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |