missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Test in process.
Specifications
Specifications
| Accession Number | Q92913 |
| Concentration | LYOPH |
| For Use With (Application) | Western Blot, PAGE |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris, pH 8.5, 500 mM NaCl. |
| Gene ID (Entrez) | 2258 |
| Molecular Weight (g/mol) | 27.6 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of FGF-13 as determined by LAL method. |
| Gene Alias | FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?