missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FGF-12 Protein

Product Code. 18319227
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA.
Unit Size:
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18319227 Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA. 25µg
18186350 Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA with BSA as a carrier protein. 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18319227 Supplier R&D Systems™ Supplier No. 2246FG025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. When Recombinant Human FGFR4 Fc Chimera (Catalog # 685-FR ) is immobilized at 2 μg/mL, 100 μL/well, Recombinant FGF-12 binds with an ED50 of 3-18 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_004104
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA.
Gene ID (Entrez) 2257
Molecular Weight (g/mol) M.W. (theoretical): 20.4 kDa
Name FGF-12
Quantity 25 μg
Source E. coli-derived human FGF-12 protein Met1-Thr181
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol FGF12
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein FGF-12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.