missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FGF-10 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT.
Unit Size:
25µg
Description
The Recombinant Human FGF-10 Protein is derived from E. coli. The Recombinant Human FGF-10 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT. |
| Gene ID (Entrez) | 2255 |
| Molecular Weight (g/mol) | Observed Molecular Wt.: 19-22 kDa, reducing conditions |
| Name | FGF-10 |
| Quantity | 25 μg |
| Source | E. coli-derived human FGF-10 protein Cys37-Ser208 & Gly41-Ser208 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | FGF10, FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, KGF2, KGF-2, produced by fibroblasts of urinary bladder lamina propria |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction