missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ferritin Protein

A cDNA sequence encoding the Ferritin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11867-EC-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15938638

  • £3965.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Ferritin Protein
Research Use Only
Recombinant Protein
E. coli
The protein solution is in 0.05M TRIS buffer pH 7.5 containing 1.0M NaCl and 0.09% NaN3.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Greater than 96.0%.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers