missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Feline CXCL12/SDF-1 beta (aa 22-93)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 351-FS-050/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18388937

  • £394.16 / 50µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human/Feline CXCL12/SDF-1 beta (aa 22-93) is derived from E. coli. The Recombinant Human/Feline CXCL12/SDF-1 beta (aa 22-93) has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
CXCL12/SDF-1 beta aa 22-93
E. coli-derived CXCL12/SDF-1 beta protein Lys22-Met93
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
8 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers