missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FCAR Protein
A cDNA sequence encoding the FCAR was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11857-50ug
Additional Details : Weight : 0.01000kg
Specifications
FCAR Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPQEGDFPM PFISAKSSPV IPLDGSVKIQ CQAIREAYLT QLMIIKNSTY REIGRRLKFW NETDPEFVID HMDANKAGRY QCQYRIGHYR FRYSDTLELV VTGLYGKPFL SADRGLVLMP GENISLTCSS AHIPFDRFSL AKEGELSLPQ HQSGEHPANF SLGPVDLNVS GIYRCYGWYN RSPYLWSFPS NALELVVTDS IHQDYTTQNH HHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FCAR protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |