missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fc receptor-like protein 1

A DNA sequence encoding the Homo sapiens (Human) Fc receptor-like protein 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7695-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16080241

  • £2065.00 / 1mg
Estimated Shipment: 15-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001004310.2
Fc receptor-like protein 1
Research Use Only
FCRL6
Recombinant Protein
E. coli
LYLQAWPNPV FEGDALTLRC QGWKNTPLSQ VKFYRDGKFL HFSKENQTLS MGAATVQSRG QYSCSGQVMY IPQTFTQTSE TAMVQVQELF PPPVLSAIPS PEPREGSLVT LRCQTKLHPL RSALRLLFSF HKDGHTLQDR GPHPELCIPG AKEGDSGLYW CEVAPEGGQV QKQSPQLEVR VQAPVSRPVL TLHHGPADPA VGDMVQLLCE AQRGSPPILY SFYLDEKIVG NHSAPCGGTT SLLFPVKSEQ DAGNYSCEAE NSVSRERSEP KKLSLKGSQV LFTPASNW
Greater than 90% as determined by SDS-PAGE.
343413
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers