missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Fc gamma RIIIB/CD16b/SH His Protein, CF
Description
Measured by its binding ability in a functional ELISA. Recombinant Human Fc gamma RIIIB/CD16b/SH His-tag (Catalog # 11709-FC) binds Human IgG with an ED50 of 0.100-1.50 Âμg/mL.
Specifications
Specifications
| Accession Number | O75015.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 2215 |
| Molecular Weight (g/mol) | M.W.-observed: 36-54 kDa, under reducing conditions., M.W.-theroretical: 22 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human Fc gamma RIIIB/CD16b protein Gly17-Ser200, with a C-terminal 6-His tag Asn65Ser, Ala78Asp |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD16, CD16b, CD16b antigen, Fc fragment of IgG, low affinity IIIb, receptor (CD16b), Fc fragment of IgG, low affinity IIIb, receptor for (CD16), FCG3, Fc-gamma receptor IIIb (CD 16), Fc-gamma RIII, Fc-gamma RIIIb, Fc-gamma RIII-beta, FCGR3, FCGR3B, FcgRIIIB, FcR-10, fcRIII, fcRIIIb, IGFR3, IgG Fc receptor III-1, low affinity immunoglobulin gamma Fc region receptor III-B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?