missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Fc gamma RIIIB/CD16b/NA1 His Protein, CF

Product Code. 30689928
Change view
Click to view available options
:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30689928 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689928 Supplier R&D Systems™ Supplier No. 11652FC050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. Recombinant Human Fc gamma RIIIB/CD16b/NA1 His-tag (Catalog # 11652-FC) binds Human IgG with an ED50 of 0.100-1.50 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75015.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 2215
Molecular Weight (g/mol) M.W.-observed: 33-45 kDa, under reducing conditions., M.W.-theroretical: 22 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Fc gamma RIIIB/CD16b protein Gly17-Ser200, with a C-terminal 6-His tag Ser36Arg, Asn82Asp, Ile106Val
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CD16, CD16b, CD16b antigen, Fc fragment of IgG, low affinity IIIb, receptor (CD16b), Fc fragment of IgG, low affinity IIIb, receptor for (CD16), FCG3, Fc-gamma receptor IIIb (CD 16), Fc-gamma RIII, Fc-gamma RIIIb, Fc-gamma RIII-beta, FCGR3, FCGR3B, FcgRIIIB, FcR-10, fcRIII, fcRIIIb, IGFR3, IgG Fc receptor III-1, low affinity immunoglobulin gamma Fc region receptor III-B
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in water.
Protein Fc gamma RIIIB/CD16b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.