missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ Recombinant Human Fc gamma RIIIB/CD16b His Avi Protein, CF

Product Code. 18794444
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18794444 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18794444 Supplier R&DSystems™ Supplier No. AVI11507050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

His-tag Avi-tag

Measured by its binding ability in a functional ELISA. Recombinant Human Fc gamma RIIIB/CD16b His-tag Avi-tag (Catalog # AVI11507) binds Human IgG with an ED50 of 60.0-720 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75015.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 2215
Molecular Weight (g/mol) M.W.-observed: 39-63 kDa, under reducing conditions., M.W.-theroretical: 24 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Fc gamma RIIIB/CD16b protein Human FCGRIIIB (Gly17-Gln208) (N-terminus) Accession # O75015.2 6-His tag Avi-tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CD16, CD16b, CD16b antigen, Fc fragment of IgG, low affinity IIIb, receptor (CD16b), Fc fragment of IgG, low affinity IIIb, receptor for (CD16), FCG3, Fc-gamma receptor IIIb (CD 16), Fc-gamma RIII, Fc-gamma RIIIb, Fc-gamma RIII-beta, FCGR3, FCGR3B, FcgRIIIB, FcR-10, fcRIII, fcRIIIb, IGFR3, IgG Fc receptor III-1, low affinity immunoglobulin gamma Fc region receptor III-B
Conjugate Biotin
Reconstitution Reconstitute at 500 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Fc gamma RIIIB/CD16b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.