missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Fc gamma RIIA/CD32a (R167) Protein

Product Code. 18735359
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18735359

Brand: R&D Systems™ 1330CD01M/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Human Fc gamma RIIA/CD32a (R167) Protein is derived from NS0. The Recombinant Human Fc gamma RIIA/CD32a (R167) Protein has been validated for the following applications: Binding Activity.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAA35827
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 2212
Molecular Weight (g/mol) Observed M.W.: 26-36 kDa, under reducing conditions.
Quantity 1 mg
Source Mouse myeloma cell line,NS0-derived human Fc gamma RIIA/CD32a protein Ala36-Ile218,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias CD32 antigen, CD32A, CD32MGC23887
Gene Symbol FCGR2A
Biological Activity <80nM
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.