missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Fc epsilon RI alpha Protein
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human Fc epsilon RI alpha is immobilized at 0.1 μg/mL (100 μL/well), the concentration of human IgE that produces 50% of the optimal binding response is found to be approximately 0.0200-0.200 μg/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 2205 |
| Molecular Weight (g/mol) | 21.9 kDa |
| Name | Fc epsilon RI alpha |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Fc epsilon RI alpha protein Val26-Gln205,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?