missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAM3A Protein
A cDNA sequence encoding the FAM3A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11844-1mg
Additional Details : Weight : 0.01000kg
Specifications
FAM3A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASFPRIQQLFTS PESSVTAAPR ARKYKCGLPQ PCPEEHLAFR VVSGAANVIG PKICLEDKML MSSVKDNVGR GLNIALVNGV SGELIEARAF DMWAGDVNDL LKFIRPLHEG TLVFVASYDD PATKMNEETR KLFSELGSRN AKELAFRDSW VFVGAKGVQN KSPFEQHVKN SKHSNKYEGW PEALEMEGCI PRRSTAS | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FAM3A filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 20mM TRIS and 50mM NaCl, pH 7.5. |