missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAM19A5 Protein
A cDNA sequence encoding the FAM19A5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11843-10ug
Additional Details : Weight : 0.01000kg
Specifications
25817 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASQFLKEGQLAA GTCEIVTLDR DSSQPRRTIA RQTARCACRK GQIAGTTRAR PACVDARIIK TKQWCDMLPC LEGEGCDLLI NRSGWTCTQP GGRIKTTTVS | |
Greater than 95.0% as determined by SDS-PAGE. |
FAM19A5 Protein | |
Research Use Only | |
FAM19A5 | |
Human | |
His | |
FAM19A5 filtered (0.4microm) solution at a concentration of 0.2 mg/ml in 20mM Tris buffer, 50mM NaCl, pH 7.5 and 10% (w/v) glycerol. |