missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FABP3 Protein

A cDNA sequence encoding the FABP3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11835-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928528

  • £3965.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FABP3 Protein
Research Use Only
Recombinant Protein
E. coli
MVDAFLGTWK LVDSKNFDDY MKSLGVGFAT RQVASMTKPT TIIEKNGDIL TLKTHSTFKN TEISFKLGVE FDETTADDRK VKSIVTLDGG KLVHLQKWDG QETTLVRELI DGKLILTLTH GTAVCTRTYE KEA
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
50mM phosphate borate buffer pH 8.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers