missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human F-box / LRR-repeat protein 1

Product Code. 16033571
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16033571

Brand: enQuireBio™ QP8251ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) F-box/LRR-repeat protein 2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001165184.1
Gene ID (Entrez) 25827
Name F-box / LRR-repeat protein 1
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol FBXL2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MVFSNNDEGL INKKLPKELL LRIFSFLDIV TLCRCAQISK AWNILALDGS NWQRIDLFNF QTDVEGRVVE NISKRCGGFL RKLSLRGCIG VGDSSLKTFA QNCRNIEHLN LNGCTKITDS TCYSLSRFCS KLKHLDLTSC VSITNSSLKG ISEGCRNLEY LNLSWCDQIT KDGIEALVRG CRGLKALLLR GCTQLEDEAL KHIQNYCHEL VSLNLQSCSR ITDEGVVQIC RGCHRLQALC LSGCSNLTDA SLTALGLNCP RLQILEAARC SHLTDAGFTL LARNCHELEK MDLEECILIT DSTLIQLSIH CPKLQALSLS HCELITDDGI LHLSNSTCGH ERLRVLELDN CLLITDVALE HLENCRGLER LELYDCQQVT RAGIKRMRAQ LPHVKVHAYF APVTPPTAVA GSGQRLCRCC VIL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.