missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F-actin-capping protein subunit alpha-2 Protein
A DNA sequence encoding the Homo sapiens (Human) F-actin-capping protein subunit alpha-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5759-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006127.1 | |
F-actin-capping protein subunit alpha-2 Protein | |
Research Use Only | |
CAPZA2 | |
Recombinant Protein | |
E. coli | |
ADLEEQLSDE EKVRIAAKFI IHAPPGEFNE VFNDVRLLLN NDNLLREGAA HAFAQYNLDQ FTPVKIEGYE DQVLITEHGD LGNGKFLDPK NRICFKFDHL RKEATDPRPC EVENAVESWR TSVETALRAY VKEHYPNGVC TVYGKKIDGQ QTIIACIESH QFQAKNFWNG RWRSEWKFTI TPSTTQVVGI LKIQVHYYED GNVQLVSHKD IQDSLTVSNE VQTAKEFIKI VEAAENEYQT AISENYQTMS DTTFKALRRQ LPVTRTKIDW NKILSYKIGK EMQNA | |
Greater than 90% as determined by SDS-PAGE. |
830 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |