missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EXOSC7 Protein

Product Code. 15958488
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15958488

Brand: enQuireBio™ QP118225ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the EXOSC7 was constructed and used to recombinantly synthesize the protein.

Specifications

Name EXOSC7 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMASVTLS EAEKVYIVHG VQEDLRVDGR GCEDYRCVEV ETDVVSNTSG SARVKLGHTD ILVGVKAEMG TPKLEKPNEG YLEFFVDCSA SATPEFEGRG GDDLGTEIAN TLYRIFNNKS SVDLKTLCIS PREHCWVLYV DVLLLECGGN LFDAISIAVK AALFNTRIPR VRVLEDEEGS KDIELSDDPY DCIRLSVENV PCIVTLCKIG YRHVVDATLQ EEACSLASLL VSVTSKGVVT CMRKVGKGSL DPESIFEMME TGKRVGKVLH ASLQSVVHKE ESLGPKRQKV GFLG
Buffer EXOSC7 protein solution (0.5 mg/ml) containing 20mM Phosphate buffer saline (pH 8.0), 0.15M NaCl, 1mM DTT and 20% glycerol.
Purity or Quality Grade Greater than 85.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.