missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ETS2 Protein
A cDNA sequence encoding the ETS2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | ETS2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948468
|
enQuireBio™
QP11815-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938468
|
enQuireBio™
QP11815-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928468
|
enQuireBio™
QP11815-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ETS2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ETS2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MNDFGIKNMD QVAPVANSYR GTLKRQPAFD TFDGSLFAVF PSLNEEQTLQ EVPTGLDSIS HDSANCELPL LTPCSKAVMS QALKATFSGF KKEQRRLGIP KNPWLWSEQQ VCQWLLWATN EFSLVNVNLQ RFGMNGQMLC NLGKERFLEL APDFVGDILW EHLEQMIKEN QEKTEDQYEE NSHLTSVPHW INSNTLGFGT EQAPYGMQTQ NYPKGGLLDS MCPASTPSVL SSEQEFQMFP KSRLSSVSVT YCSVSQDFPG SNLNLLTNNS GTPKDHDSPE NGADSFESSD SLLQSWNSQS SLLDVQRVPS FESFEDDCSQ SLCLNKPTMS FKDYIQERSD PVEQGKPVIP AAVLAGFTGS GPIQLWQFLL ELLSDKSCQS FISWTGDGWE FKLADPDEVA RRWGKRKNKP KMNYEKLSRG LRYYYDKNII HKTSGKRYVY RFVCDLQNLL GFTPEELHAI LGVQPDTED | |
Greater than 85.0% as determined by SDS-PAGE. |