missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ETS1 Protein

Product Code. 15908468
Click to view available options
Quantity:
10 μg
2 μg
5 μg
Unit Size:
10µg
2µg
5µg
This item is not returnable. View return policy

Product Code. 15908468

Brand: enQuireBio™ QP118142ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the ETS1 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 2113
Name ETS1 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol ETS1
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MKAAVDLKPT LTIIKTEKVD LELFPSPDME CADVPLLTPS SKEMMSQALK ATFSGFTKEQ QRLGIPKDPR QWTETHVRDW VMWAVNEFSL KGVDFQKFCM NGAALCALGK DCFLELAPDF VGDILWEHLE ILQKEDVKPY QVNGVNPAYP ESRYTSDYFI SYGIEHAQCV PPSEFSEPSF ITESYQTLHP ISSEELLSLK YENDYPSVIL RDPLQTDTLQ NDYFAIKQEV VTPDNMCMGR TSRGKLGGQD SFESIESYDS CDRLTQSWSS QSSFNSLQRV PSYDSFDSED YPAALPNHKP KGTFKDYVRD RADLNKDKPV IPAAALAGYT GSGPIQLWQF LLELLTDKSC QSFISWTGDG WEFKLSDPDE VARRWGKRKN KPKMNYEKLS RGLRYYYDKN IIHKTAGKRY VYRFVCDLQS LLGYTPEELH AMLDVKPDAD E
Buffer ETS1 protein in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.